Order Entry
Canada
ContactUsLinkComponent
Human Periostin - OSF2 ELISA Kit, Rockland Immunochemicals
Human Periostin - OSF2 ELISA Kit, Rockland Immunochemicals
Catalog # 76235-748
CAS Number:  
Human Periostin - OSF2 ELISA Kit, Rockland Immunochemicals
Catalog # 76235-748
Supplier Number:  KOA0613
CAS Number:  

Specifications

  • Primary antibody reactivity:
    Human
  • Description:
    Human Periostin - OSF2 ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76235-748

Specifications

About this item

Human Periostin - OSF2 AccuSignal ELISA Kit

  • Natural and recombinant human Periostin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: N22-Q836

Periostin, also known as OSF2, is a protein that in humans is encoded by the POSTN gene. The International Radiation Hybrid Mapping Consortium mapped the POSTN gene to chromosome 13. Periostin functions as a ligand for alpha-V/beta-3 and alpha-V/beta-5 integrins to support adhesion and migration of epithelial cells. It is found that periostin was overexpressed by the majority of human primary breast cancers examined. After myocardial infarction, periostin-induced cardiomyocyte cell cycle reentry and mitosis were associated with improved ventricular remodeling and myocardial function, reduced fibrosis and infarct size, and increase angiogenesis.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human Periostin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human Periostin standard solution and each sample be measured in duplicate.