Specifications
- Assay Type:Sandwich
- Primary antibody reactivity:Mouse
- Description:Mouse DKK1 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76235-686
Specifications
About this item
Mouse DKK1 AccuSignal ELISA Kit
- Natural and recombinant mouse DKK1.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: CHO; Immunogen sequence: S30-H272
Dickkopf-related protein 1 is a protein that in humans is encoded by the DKK1 gene, which was mapped to chromosome 10q11.2. This gene encodes a protein that is a member of the dickkopf family. It is a secreted protein with two cysteine rich regions and is involved in embryonic development through its inhibition of the WNT signaling pathway. Elevated levels of DKK1 in bone marrow, plasma and peripheral blood is associated with the presence of osteolytic bone lesions in patients with multiple myeloma.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 3000pg/ml, 1500pg/ml, 750pg/ml, 419.pg/ml, 187.5pg/ml, 93.8pg/ml, 46.9pg/ml mouse DKK1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernatants, serum or plasma (heparin, EDTA) to each empty well. We recommend that each mouse DKK1 standard solution and each sample is measured in duplicate.