Order Entry
Canada
ContactUsLinkComponent
Human KLK8 - Kallikrein-8 ELISA Kit, Rockland Immunochemicals
Human KLK8 - Kallikrein-8 ELISA Kit, Rockland Immunochemicals
Catalog # 76237-618
CAS Number:  
Human KLK8 - Kallikrein-8 ELISA Kit, Rockland Immunochemicals
Catalog # 76237-618
Supplier Number:  KOA0489
CAS Number:  

Specifications

  • Primary antibody reactivity:
    Human
  • Description:
    Human KLK8 - Kallikrein-8 ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76237-618

Specifications

About this item

Human KLK8 - Kallikrein-8 AccuSignal ELISA Kit

  • Natural and recombinant human Kallikrein 8.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: Q29-G260

Kallikrein-8 is a protein that in humans is encoded by the KLK8 gene. Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in a cluster on chromosome 19. Alternate splicing of this gene results in four transcript variants encoding four different isoforms. The isoforms exhibit distinct patterns of expression that suggest roles in brain plasticity and ovarian cancer.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human KLK8 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, cell lysates, tissue homogenates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human KLK8 standard solution and each sample be measured in duplicate.