- Primary antibody reactivity:Human
- Description:Human CCL13 - MCP4 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76236-904
Human CCL13 - MCP4 AccuSignal ELISA Kit
- Natural and recombinant human CCL13.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: E.coli; Immunogen sequence: Q24-T98
Chemokine(C-C motif) ligand 13(CCL13) is a small cytokine belonging to the CC chemokine family. Its gene is located on human chromosome 17 within a large cluster of other CC chemokines. CCL13 induces chemotaxis in monocytes, eosinophils, T lymphocytes, and basophils by binding cell surface G-protein linked chemokine receptors such as CCR2, CCR3 and CCR5. Activity of this chemokine has been implicated in allergic reactions such as asthma. CCL13 can be induced by the inflammatory cytokines interleukin-1 and TNF-alpha.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.3pg/ml, 15.6pg/ml, 7.8pg/ml human CCL13 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. We recommend that each human CCL13 standard solution and each sample is measured in duplicate.