Order Entry
Canada
ContactUsLinkComponent
Mouse IL-27 p28 ELISA Kit, Rockland Immunochemicals
Mouse IL-27 p28 ELISA Kit, Rockland Immunochemicals
  76237-330
 :  
Mouse IL-27 p28 ELISA Kit, Rockland Immunochemicals
  76237-330
 :  KOA0473
 :  

 

  • Primary antibody reactivity:
    Mouse
  • Target protein:
    IL-27 p28
  • Description:
    Mouse IL-27 p28 ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76237-330

 

 

Mouse Interleukin-27 p28 AccuSignal ELISA Kit

  • Natural and recombinant mouse IL-27 p28.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: F29-S234

Interleukin-27(IL-27) is a heterodimeric cytokine belonging to the IL-12 family that is composed of two subunits; Epstein-Barr virus(EBV)-induced gene 3(EBI3)(also known as IL-27B) and IL27-p28(known as IL-30). IL-27 is produced by antigen-presenting cells. By genomic sequence analysis, the IL27 p28 gene was mapped to chromosome 16p11. L-27 plays an important function in regulating the activity of B- and T-lymphocytes. The effects of IL-27 are eliciting by its interaction with a specific cell-surface receptor complex composed of two proteins known as IL27R and gp130.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml mouse IL-27 p28 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse IL-27 p28 standard solution and each sample be measured in duplicate.