Order Entry
Canada
ContactUsLinkComponent
Mouse IL-7 ELISA Kit, Rockland Immunochemicals
Mouse IL-7 ELISA Kit, Rockland Immunochemicals
Catalog # 76235-638
CAS Number:  
Mouse IL-7 ELISA Kit, Rockland Immunochemicals
Catalog # 76235-638
Supplier Number:  KOA0460
CAS Number:  

Specifications

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse IL-7 ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76235-638

Specifications

About this item

Mouse Interleukin-7 AccuSignal ELISA Kit

  • Natural and recombinant mouse IL-7.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.colI; Immunogen sequence: E26-I154

Interleukin 7(IL7) is a protein that is encoded by the IL7 gene in humans. The gene for human interleukin 7(IL7) maps to chromosome 8 by Southern analysis of a somatic cell hybrid panel and to 8q12-q13 by in situ hybridization.1IL-7 is essential for survival and expansion of pathogenic T helper type 17(T(H)17) cells in experimental autoimmune encephalomyelitis(EAE).2

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml mouse IL-7 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(EDTA) to each empty well. It is recommended that each mouse IL-7 standard solution and each sample be measured in duplicate.