Order Entry
Canada
ContactUsLinkComponent
Mouse TNFRSF18 - GITR ELISA Kit, Rockland Immunochemicals
Mouse TNFRSF18 - GITR ELISA Kit, Rockland Immunochemicals
Catalog # 76235-628
CAS Number:  
Mouse TNFRSF18 - GITR ELISA Kit, Rockland Immunochemicals
Catalog # 76235-628
Supplier Number:  KOA0455
CAS Number:  

Specifications

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse TNFRSF18 - GITR ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76235-628

Specifications

About this item

Mouse TNFRSF18 - GITR AccuSignal ELISA Kit

  • Natural and recombinant mouse TNFRSF18.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: S22-H153

Tumor necrosis factor receptor superfamily member 18(TNFRSF18), also called GITR or AITR is a protein that in humans is encoded by the TNFRSF18 gene. This gene is mapped to 1p36.33. This gene encodes a member of the TNF-receptor superfamily. The encoded receptor has been shown to have increased expression upon T-cell activation, and it is thought to play a key role in dominant immunological self-tolerance maintained by CD25(+)CD4(+) regulatory T cells. Knockout studies in mice also suggest the role of this receptor is in the regulation of CD3-driven T-cell activation and programmed cell death. Three alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml mouse TNFRSF18 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse TNFRSF18 standard solution and each sample be measured in duplicate.