Order Entry
Canada
ContactUsLinkComponent
Human ErbB-2 ELISA Kit, Rockland Immunochemicals
Human ErbB-2 ELISA Kit, Rockland Immunochemicals
Catalog # 76235-612
CAS Number:  
Human ErbB-2 ELISA Kit, Rockland Immunochemicals
Catalog # 76235-612
Supplier Number:  KOA0447
CAS Number:  

Specifications

  • Primary antibody reactivity:
    Human
  • Description:
    Human ErbB-2 ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76235-612

Specifications

About this item

Human ErbB-2 AccuSignal ELISA Kit

  • Natural and recombinant human ErbB-2.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: T23-T652

HER2/neu(also known as ErbB-2) stands for "Human Epidermal growth factor Receptor 2" and is a protein giving higher aggressiveness in breast cancers. It is a member of the ErbB protein family, more commonly known as the epidermal growth factor receptor family. HER2/neu has also been designated as CD340(cluster of differentiation 340) and p185. It is encoded by the ERBB2 gene.HER2 is a cell membrane surface-bound receptor tyrosine kinase and is normally involved in the signal transduction pathways leading to cell growth and differentiation. It is encoded within the genome by HER2/neu, a known proto-oncogene. HER2 is thought to be an orphan receptor, with none of the EGF family of ligands able to activate it. However, ErbB receptors dimerise on ligand binding, and HER2 is the preferential dimerisation partner of other members of the ErbB family.1 The HER2 gene is a proto-oncogene located at the long arm of human chromosome 17(17q21-q22)2.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human ErbB-2 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernatants, serum, plasma(heparin, EDTA) or tissue lysates to each empty well. It is recommended that each human ErbB-2 standard solution and each sample be measured in duplicate.