Specifications
- Primary antibody reactivity:Human
- Description:Human DCR3 - TNFRSF6B ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76237-306
Specifications
About this item
Human DCR3 - TNFRSF6B AccuSignal ELISA Kit
- Natural and recombinant human DCR3.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: sf21; Immunogen sequence: V24-H300
Decoy receptor 3 (Dcr3), also known as tumor necrosis factor receptor superfamily member 6B (TNFRSF6B), TR6 and M68, is a soluble protein of the tumor necrosis factor receptor superfamily which inhibits Fas ligand-inducedapoptosis. This gene belongs to the tumor necrosis factor receptor superfamily. It acts as a decoy receptor that competes with death receptors for ligand binding. The encoded protein is postulated to play a regulatory role in suppressing FasL- and LIGHT-mediated cell death and T cell activation as well as to induce angiogenesis via neutralization of TL1A. Overexpression of this gene has been noted in various tumors e.g. gastrointestinal tract tumors, and it is located in a gene-rich cluster on chromosome 20, with other potentially tumor-related genes.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human DCR3 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human DCR3 standard solution and each sample be measured in duplicate.