- Assay Type:Sandwich
- Description:Rat CD80 - B7-1 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76236-890
Rat CD80 - B7-1 AccuSignal ELISA Kit
- Natural and recombinant rat CD80.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: I39-Q248
The protein CD80(Cluster of Differentiation 80) is a molecule found on activated B cells and monocytes which provides a costimulatory signal necessary for T cell activation and survival. It is also known as B7-1. The cDNA for B7-1 predicts a type I membrane protein, i.e., one synthesized with a signal peptide that is cleaved upon translocation across the endoplasmic membrane. The protein is predicted to contain 2 extracellular domains structurally similar to those of Ig, a hydrophobic transmembrane region, and a short cytoplasmic domain.The CD80 and CD86 genes encode B7-1 and B7-2, respectively, which are structurally similar members of the immunoglobulin superfamily expressed on a variety of hematopoietic cell types. B7-1 and B7-2 provide a costimulatory signal to T cells by interacting with CD28 and CTLA4.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml rat CD80 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernates or serum to each empty well. It is recommended that each rat CD80 standard solution and each sample be measured in duplicate.