Order Entry
Canada
ContactUsLinkComponent
Human CD56 - NCAM-1 ELISA Kit, Rockland Immunochemicals
Human CD56 - NCAM-1 ELISA Kit, Rockland Immunochemicals
Catalog # 76236-884
CAS Number:  
Human CD56 - NCAM-1 ELISA Kit, Rockland Immunochemicals
Catalog # 76236-884
Supplier Number:  KOA0409
CAS Number:  

Specifications

  • Primary antibody reactivity:
    Human
  • Description:
    Human CD56 - NCAM-1 ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76236-884

Specifications

About this item

Human CD56 - NCAM-1 AccuSignal ELISA Kit

  • Natural and recombinant human CD56.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: L20-G718

Neural Cell Adhesion Molecule, NCAM, also known as the cluster of differentiation CD56, is a hemophilic binding glycoprotein. It is a glycoprotein of Immunoglobulin(Ig) super family. At least 27 alternatively spliced NCAM mRNAs are produced, giving a wide diversity of NCAM isoforms NCAM gene is located at 11q22-q23. This glycoprotein is mainly expressed on the surface of neurons, glia, skeletal muscle and natural killer cells. NCAM has been implicated as having a role in cell–cell adhesion, neurite outgrowth, synaptic plasticity, and learning and memory.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human CD56 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. We recommend that each human CD56 standard solution and each sample is measured in duplicate.