- Primary antibody reactivity:Mouse
- Target protein:CD40
- Description:Mouse CD40 - TNFRSF5 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76237-274
Mouse CD40 - TNFRSF5 AccuSignal ELISA Kit
- Natural and recombinant mouse CD40.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: V24-R193
CD40 is a cell surface receptor that is expressed on the surface of all mature B cells, most mature B-cell malignancies, and some early B-cell acute lymphocytic leukemias, but is not expressed on plasma cells. The CD40 gene is localized to the long arm of chromosome 20, bands q12-q13.2. The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor has been found to be essential in mediating a broad variety of immune and inflammatory responses including T cell-dependent immunoglobulin class switching, memory B cell development, and germinal center formation.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml mouse CD40 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, cell lysates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse CD40 standard solution and each sample be measured in duplicate.