- Primary antibody reactivity:Human
- Description:Human TNF beta ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76236-862
Human TNF beta AccuSignal ELISA Kit
- Natural and recombinant human TNF beta.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: E.coli; Immunogen sequence: L35-L205
Tumor necrosis factor-beta(TNF-beta), previously called lymphotoxin-alpha(LTA), is a cytokine produced by lymphocytes. TNF-alpha and TNF-beta share 35% identity and 50% homology in the amino acid sequence. The substance LTA mediates a wide variety of inflammatory, immunostimulatory, and antiviral responses. LTA is associated with susceptibility to myocardial infarction, asthma and other diseases. The LTA gene is located on human chromosome 6. The standard product used in this kit is recombinant protein, consisting of 172 amino acids with the molecular mass of 18.8KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human TNF beta standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA, citrate) to each empty well. It is recommended that each human TNF beta standard solution and each sample be measured in duplicate.