Order Entry
Canada
ContactUsLinkComponent
Mouse VEGFR3 - FLT4 ELISA Kit, Rockland Immunochemicals
Mouse VEGFR3 - FLT4 ELISA Kit, Rockland Immunochemicals
Catalog # 76236-830
CAS Number:  
Mouse VEGFR3 - FLT4 ELISA Kit, Rockland Immunochemicals
Catalog # 76236-830
Supplier Number:  KOA0334
CAS Number:  

Specifications

  • Primary antibody reactivity:
    Mouse
  • Target protein:
    VEGF-R3/FLT-4
  • Description:
    Mouse VEGFR3 - FLT4 ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76236-830

Specifications

About this item

Mouse VEGFR3 - FLT4 AccuSignal ELISA Kit

  • Natural and recombinant mouse VEGFR3.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: Y25‑D770

Fms-related tyrosine kinase 4, also known as FLT4 or VEGFR3, is a protein which in humans is encoded by the FLT4 gene. It is mapped to 5q35.3. This gene encodes a tyrosine kinase receptor for vascular endothelial growth factors C and D. The protein is thought to be involved in lymphangiogenesis and maintenance of the lymphatic endothelium. FLT4 has an essential role in the development of the embryonic cardiovascular system before the emergence of the lymphatic vessels. It has been found that FLT4, which provides proangiogenic signaling when expressed on endothelium, may also have antiangiogenic properties when expressed at an avascular site by nonendothelial cells. FLT4 is also regarded as a regulator of vascular network formation.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml mouse VEGFR3 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates or serum to each empty well. It is recommended that each mouse VEGFR3 standard solution and each sample be measured in duplicate.