- Primary antibody reactivity:Human
- Description:Human TIE2 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76236-784
Human TIE2 AccuSignal ELISA Kit
- Natural and recombinant human TIE2.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: A23-K745
Tyrosine kinase with Ig and EGF homology domain 2(Tie-2), also called TEK tyrosine kinase, endothelial(TEK). Tie-2 and tie-1 are expressed in early embryonic vascular system and in maternal decidual vascular endothelial cells, where the vasculature undergoes an active angiogenesis. Tie-2, but not tie-1, expression was also detected in extraembryonic mesoderm of the amnion. Angiogenesis is coordinated with follicular cell growth in goitrogenesis. The angiopoietins, Ang-1 and Ang-2, are angiogenic growth factors acting through Tie-2. Tie-2 and Ang-1 are expressed in thyroid epithelial and endothelial cells, and Tie-2 is regulated by TSH and cAMP in follicular cells. And Tie-2 expression is increased in goiter in both humans and rats, consistent with a role in goitrogenesis. Tie2/Ang-1 signaling pathway plays a critical role in the maintenance of HSCs in a quiescent state in the BM niche. And the Tie-2 signaling pathway is also critical for endothelial cell-smooth muscle cell communication in venous morphogenesi...
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human TIE2 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human TIE2 standard solution and each sample be measured in duplicate.