Order Entry
Canada
ContactUsLinkComponent
Human MMP-9 ELISA Kit, Rockland Immunochemicals
Human MMP-9 ELISA Kit, Rockland Immunochemicals
Catalog # 76236-750
CAS Number:  
Human MMP-9 ELISA Kit, Rockland Immunochemicals
Catalog # 76236-750
Supplier Number:  KOA0273
CAS Number:  

Specifications

  • Primary antibody reactivity:
    Human
  • Target protein:
    MMP-9
  • Description:
    Human MMP-9 ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76236-750

Specifications

About this item

Human MMP-9 AccuSignal ELISA Kit

  • Natural and recombinant human total MMP-9.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence:    A20-D707

The 92-kD type IV collagenase is also known as 92-kD gelatinase, type V collagenase, gelatinase B, or matrix metalloproteinase-9(MMP9). The 72- and 92-kDa type IV collagenases are members of a group of secreted zinc metalloproteases. The matrix metalloproteinases(MMPs) are able to degrade the extracellular matrix and allow angiogenesis and tumor invasion. Gelatinase B, a matrix metalloproteinase that has proteolytic activity against connective tissue proteins, has been suggested to be important in the connective tissue remodeling processes associated with atherogenesis and plaque rupture. MMP-9 is predominantly expressed in neutrophils, macrophages, and mast cells, rather than in oncogene-positive neoplastic cells. The polymorphism of MMP-9 acts as a genetic factor for the development of smoking-induced pulmonary emphysema. The standard product used in this kit is recombinant human MMP-9 with the molecular mass of 95KDa. The detected MMP-9 includes zymogen and active enzyme.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human MMP-9 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma(heparin), saliva or urine to each empty well. It is recommended that each human MMP-9 standard solution and each sample be measured in duplicate.