- Assay Type:Sandwich
- Primary antibody reactivity:Mouse
- Description:Mouse Laminin ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76236-706
Mouse Laminin AccuSignal ELISA Kit
- Natural mouse Laminin.
- There is no detectable cross-reactivity with other relevant proteins.
- From murine sarcoma basement membrane
Laminin is a large basement membrane glycoprotein composed of three subunits designated the A, B1, and B2. Laminin has diverse biological functions, which include stimulating epithelial cell growth and differentiation. The nucleotide sequence of human laminin A chain has an open reading frame encoding 3075-amino acids. The human laminin A chain is at locus 18p11.3. The nucleotide sequence of the human laminin B1 reveals a 5358-base pair open reading frame that potentially codes for 1786 amino acids, including 20 amino acids of a presumptive signal peptide. The gene for the human laminin-B1 chain has been localized to chromosome 7, band q31. The B2 chain consists of six distinct domains, including two domains with alpha-helical, coiled-coil structures, two domains with cysteine-rich homologous repeats, and two globular domains. The amino acid sequences of the B2 and B1 chains demonstrate considerable homology.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml mouse Laminin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse Laminin standard solution and each sample be measured in duplicate.