Order Entry
Canada
ContactUsLinkComponent
Mouse IL-4 ELISA Kit, Rockland Immunochemicals
Mouse IL-4 ELISA Kit, Rockland Immunochemicals
Catalog # 76236-666
CAS Number:  
Mouse IL-4 ELISA Kit, Rockland Immunochemicals
Catalog # 76236-666
Supplier Number:  KOA0221
CAS Number:  

Specifications

  • Primary antibody reactivity:
    Mouse
  • Target protein:
    IL-4
  • Description:
    Mouse IL-4 ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76236-666

Specifications

About this item

Mouse Interleukin-4 AccuSignal ELISA Kit

  • Natural and recombinant mouse IL-4.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: H21-S140

Interleukin-4(IL-4), also knowns as a B-cell stimulatory factor1(BSF1), is an immunomodulatory cytokine, which can inhibit the growth of tumour cells. The human cDNA contains a single open reading frame encoding a protein of 153 amino acids, including a putative signal peptide. IL-4 may act as an autocrine growth factor in pancreatic cancer cells and also give rise to the possibility that cancer-derived IL-4 may suppress cancer-directed immunosurveillance in vivo in addition to its growth-promoting effects, thereby facilitating pancreatic tumor growth and metastasis.1 The mouse and human genes and their protein products show structural and functional similarities. The human IL-4 gene, which occurs as a single copy in the haploid genome, is mapped on chromosome 5.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.3pg/ml, 15.6pg/ml, 7.8pg/ml mouse IL-4 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates or serum to each empty well. We recommend that each mouse IL-4 standard solution and each sample is measured in duplicate.