Specifications
- Assay Type:Sandwich
- Primary antibody reactivity:Mouse
- Target protein:IGF-1
- Description:Mouse IGF-1 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76237-144
Specifications
About this item
Mouse IGF-1 AccuSignal ELISA Kit
- Natural and recombinant mouse IGF-1.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: E.coli; Immunogen sequence: G49-A118
Insulin-like growth factor 1(IGF-1) that was once called somatomedin C, is a polypeptide protein hormone similar in molecular structure to insulin. It plays an important role in childhood growth and continues to have anabolic effects in adults. Human IGF1 is a single chain 70-amino acid polypeptide cross-linked by 3 disulfide bridges, with a calculated molecular mass of 7.6 kD.1 The IGF1 gene, mapped on 12q22-q24. contains 5 exons. Exons 1-4 encode the 195-amino acid precursor(IGF1B), and exons 1, 2, 3, and 5 encode the 153-residue peptide(IGF1A). The structure of IGF1 resembles that of IGF2. And the IGF1 and IGF2 genes have complex structures with multiple promoters. The expression of both genes is regulated at the levels of transcription, RNA processing, and translation. IGF-1 is produced primarily by the liver as an endocrine hormone as well as in target tissues in a paracrine/autocrine fashion. Moreover, approximately 98% of IGF-1 is always bound to one of 6 binding proteins(IGF-BP). Furthermore, IGF-1 is one of the most potent natural activators of the AKT signaling pathway, a stimulator of cell growth and multiplication and a potent inhibitor of programmed cell death.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml mouse IGF-1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse IGF-1 standard solution and each sample be measured in duplicate.