Order Entry
Canada
ContactUsLinkComponent
Mouse FASL ELISA Kit, Rockland Immunochemicals
Mouse FASL ELISA Kit, Rockland Immunochemicals
  76236-600
 :  
Mouse FASL ELISA Kit, Rockland Immunochemicals
  76236-600
 :  KOA0166
 :  

 

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    FASL
  • Description:
    Mouse FASL ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76236-600

 

 

Mouse FASL AccuSignal ELISA Kit

  • Natural and recombinant mouse FAS-L.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: P132-L279

FAS Ligand(FASL) is a 40 kDa type II membrane protein belonging to the tumor necrosis factor family, which induces apoptosis by binding to its receptor, Fas. The human FasL gene consists of approximately 8.0 kb and is split into four exons. This gene consists of 281 amino acids with a calculated M(r) of 31,759 and was mapped on chromosome 1q23. It has an identity of 76.9% at the amino acid sequence level with mouse FasL. The FAS and FASL system plays a key role in regulating apoptotic cell death and corruption of this signalling pathway has been shown to participate in immune escape and tumorigenesis. FAS and FASL triggered apoptosis pathway plays an important role in human carcinogenesis. This system may also play a role in modulating the genetic susceptibility of mouse strains to develop T-cell lymphoblastic lymphomas.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml mouse FAS-L standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA, citrate) to each empty well. It is recommended that each mouse FAS-L standard solution and each sample be measured in duplicate.