ContactUsLinkComponent
MAPK1 Blocking Peptide
MAPK1 Blocking Peptide
Catalog #: 76222-526
CAS Number:  
MAPK1 Blocking Peptide
Catalog #: 76222-526
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Size:
    50 µg
  • Protein/peptide name:
    MAPK1 Blocking Peptide
  • Shipping temperature:
    Ambient
  • Cat. no.:
    76222-526
Specifications
About this item
ERK2 C-Term Blocking Control Peptide
  • Control Peptide - Blocking Peptide
  • PCA Assays
  • ImmunoBlot
Mitogen activiated protein kinase 1, also known as MAPK1, ERK, or ERK2, is an integral component of the MAP kinase cascade that regulates cell growth and differentiation. ERK1 and ERK2 are activated by MEK1 and MEK2 in the B-raf signaling pathway resulting in its translocation to the nucleus where it phosphorylates nuclear targets. Human ERK1 and ERK2 are 84% identical in sequence and share common functionality in cells. ERK2 is ideal for investigators involved in Neuroscience, Cell Signaling and Cancer Research.
ERK2 N-term blocking peptide is suitable as a control for polyclonal or monoclonal anti-ERK2 in immunological assays.
Customers Who Bought This Also Bought