Hex Inch Drivers, Wiha Tools
Chrome-V-molybdenum super tool steel hardened, nickel-plated, 2×200 mm, T-Handle, Red
Catalog # 76250-260
Supplier: WIHA TOOLS TE
Supplier #: 33436
Jump to:
Product Details & Documents
These drivers has chamfered tip for easy insertion.
Made of chrome-V-moly
Impact resistant
Smooth edges prevent injuries and eliminate pressure marks. The Handle is molded directly onto blade.
Specifications
- Material:Chrome-V-molybdenum super tool steel hardened, nickel-plated
- Handle color:Red
- Handle:T-Handle
- Drive size:2×200 mm