ContactUsLinkComponent
Premium 3-Layer Non-Marring Heel Grounder, Menda
Premium 3-Layer Non-Marring Heel Grounder, Menda
Catalog # 76249-744
Supplier:  Menda
undefined
Premium 3-Layer Non-Marring Heel Grounder, Menda
Catalog # 76249-744
Supplier:  Menda
Supplier Number:  17252
Specifications
  • Description:
    Non-marring Heel Grounder
  • Resistor:
    1 MΩ
  • Cat. no.:
    76249-744
  • Style:
    Hook and Loop