Orders LinkContactUsLinkComponent
Venier® Go Direct™ Sensor Carts
Venier® Go Direct™ Sensor Carts
New Product
Venier® Go Direct™ Sensor Carts
New Product
Certificates
Venier® Go Direct™ Sensor Carts
Venier® Go Direct™ Sensor Carts
3 Options Available Below
Product Details & Documents
The Go Direct Sensor cart features built-in, wireless sensors for one-dimensional dynamics and kinematics experiments.
  • Encoder wheel to report position
  • 3-axis accelerometer to measure independent acceleration
  • ±50 N force sensor to measure push and pulls
  • Mass trays for changing total mass
  • Plunger for collision and impulse studies
  • Low friction wheels for uniform motion
With Go Direct Sensor cart, students can explore force, position, velocity, and acceleration directly on their Bluetooth-enabled device - no wires or additional equipment required. Each cart features built-in sensors to simplify experiment setup and allow investigations to be conducted on or off the track. Go Direct Sensor cart can be used for hands-on kinematics and dynamics demonstrations but can also be used as an individual sensor. Hang a mass and spring from the force sensor to study simple harmonic motion, or attach the cart to a turntable to study centripetal acceleration.
The Go Direct Sensor cart can be used in a variety of experiments:
Collect position, velocity, and acceleration data as a cart rolls freely up and down an incline.
Observe collisions between two carts, test for the conservation of momentum, or measure energy changes during different types of collisions.
Investigate the relationship between force, mass, and acceleration.
Examine the energies involved in simple harmonic motion.
Measure a cart’s momentum change and compare it to the impulse it receives.
Go Direct Sensor cart connects directly to your mobile device, Chromebook™, or computer using the free Graphical Analysis™ 4 app.
Go Direct Sensor cart is also available as a part of the dynamics cart and track system with Go Direct Sensor cart item# 470310-204.
Position resolution is 0.25 mm, reported by default at 1 mm steps, force range is ±50 N and accelerometer range is ±160 m/s².
Delivery information: These includes Go Direct® Sensor cart (green or yellow), hook, flat-top rubber bumper, anti-roll peg, empty tabs (4), micro USB cable.
Product Family Options
Product Information
  • Availability
    Price
  • CA470310-208
    Vernier Go Direct™ sensor cart, green
    Vernier Go Direct™ sensor cart, green
    Log in for availability
    Log in for availability
    Each
    Each - $411.09
    $411.09
    Each
       $411.09
    Minimum order quantity is 1
    Supplier Number: GDX-CART-G
    Specifications:
    New Product
    New Product
  • CA470310-210
    Vernier Go Direct™ sensor cart, yellow
    Vernier Go Direct™ sensor cart, yellow
    Log in for availability
    Log in for availability
    Each
    Each - $411.09
    $411.09
    Each
       $411.09
    Minimum order quantity is 1
    Supplier Number: GDX-CART-Y
    Specifications:
    New Product
    New Product
  • CA470319-996
    Accessory kit for sensor carts
    Accessory kit for sensor carts
    Log in for availability
    Log in for availability
    Each
    Each - $124.09
    $124.09
    Each
       $124.09
    Minimum order quantity is 1
    Supplier Number: GDX-CART-AK
    Specifications:
    New Product
    New Product
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • 125 g masses for Vernier dynamics carts, pack of 4
    125 g masses for Vernier dynamics carts, pack of 4
    CA470303-824
    Log in for availability
    Each - $29.09