Speci-Gard® Specimen Transport Bag with Document Pouch
10153-138
: Minigrip
- Color:Transparent
- Description:Specimen transport bag with document pouch
- Dimensions:20.32×25.4 cm (8×10")
- Material:Polyethylene
- Cat. no.:10153-138
- Supplier No.:95-CMS810
Double pocket bag with press and Close® liquid-tight closure.
- Separate outside pocket holds the requisition slip
- Traditional scored opening
- With printed biohazard logo
- Without graduations
Biohazard symbol and instruction for use is printed on the cover in black and red color.
Manufactured with Micro-Tex™ polyethylene, for slip-free handling.