To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Gene ID:O35228
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Size:10 µg
- Storage conditions:Store vial at 4 °C prior to restoration. Dilute only prior to immediate use. Maintain sterility. This product DOES NOT contain preservative. DO NOT VORTEX. We recommend adding a carrier protein such as HSA or BSA to 0.1% (i.e. 1.0 mg/mL) . For best results aliquot contents and freeze at –20 °C or colder. Avoid cycles of freezing and thawing. Centrifuge vial before each opening to dislodge contents from the cap and to clarify if contents are not clear after standing at room temperature.
- Endotoxin content:Measured by LAL is <0.01ng/µg or <0.1EU/µg.
- Biological activity:Assay data for mouse recombinant EBI-3 is based upon qualitative binding to anti-EBI-3 antibody.
- Reconstitution instructions:100 µL
- Protein synonyms:IL35IL-35EBI-3 cytokineInterleukin-27 subunit beta precursorEBI-3EBI3
- Protein/peptide name:Ebi3
- Purity:>90%
- Formulation:Lyophilized
- Shipping temperature:Ambient
- Supplier No.:010-001-B66
Specifications
About this item
Epstein-Barr Virus Induced Gene-3 (EBI-3), is a secreted glycoprotein belonging to the hematopoietin receptor family and related to the p40 subunit of IL-12. It was identified by its induced expression in B-lymphocytes in response to Epstein-Barr virus infection. EBI-3 forms heterodimers with p28 to form IL-27 and with p35 to form IL-35. Both IL-27 and IL-35 have anti-inflammatory and regulatory activity. Recombinant Mouse EBI is a non-glycosylated polypeptide chain consisting of 207 amino acids with a molecular weight of 22,900 Da.