Order Entry
Canada
ContactUsLinkComponent
Fixed-Angle Rotor, Titanium, Beckman Coulter®
Fixed-Angle Rotor, Titanium, Beckman Coulter®
Catalog # 103255-094
Supplier:  Beckman Coulter
Product available on GSA Advantage®
undefined
Fixed-Angle Rotor, Titanium, Beckman Coulter®
Catalog # 103255-094
Supplier:  Beckman Coulter
Supplier Number:  393490
Product available on GSA Advantage®

Specifications

  • Model:
    MLA-150
  • Capacity:
    8× 2.0 ml
  • Description:
    Fixed Angle Rotor, Titanium
  • Cat. no.:
    103255-094
  • Maximum Speed:
    150000 rpm
  • Maximum RCF:
    1003000×g
  • Supplier No.:
    393490

Specifications

About this item

MLA-150 rotor ideal for separation of subcellular particles, viruses, protein purification/isolation, DNA and RNA.