Order Entry
Canada
ContactUsLinkComponent
 
5X DNA Sequencing Dilution Buffer III Solution, Teknova
Catalog # CA100219-902
Supplier:  Teknova
CAS Number:  
5X DNA Sequencing Dilution Buffer III Solution, Teknova
Catalog # CA100219-902
Supplier:  Teknova
Supplier Number:  D1301
CAS Number:  

Specifications

  • Description:
    Dilution Buffer III Solution
  • Size:
    500 ml
  • Cat. no.:
    CA100219-902
  • pH:
    9.0
  • Supplier No.:
    D1301

Specifications

About this item

For dilution of PE/Applied BiosystemsTM DNA sequencing reactions.

  • Contains 400 mM Tris-HCl and 1 mM MgCl₂
  • Ship and store at room temperature