ContactUsLinkComponent
Chlorine Test Kit, LaMotte
Chlorine Test Kit, LaMotte
  66170-128
 :  LaMotte
Small Business Enterprise
 :  
Chlorine Test Kit, LaMotte
  66170-128
 :  LaMotte
 :  3308-01
Small Business Enterprise
 :  
  • Description:
    DPD Free, Total and Combined Chlorine Test Kit
  • Cat. no.:
    66170-128
  • Range:
    0.2 - 3.0 ppm Cl
  • Supplier No.:
    3308
  • No. of Tests:
    50
Chlorine test kit measures free, total and combined chlorine using chlorine DPD tablets and the Octa-slide 2 comparator.
  • Range/sensitivity: 0.2 to 3.0 ppm Cl
Applications include drinking water, environmental education, food and beverage, industrial water boiling and cooling, laundry and sanitation, water and wastewater.