Order Entry
Canada
ContactUsLinkComponent
Mouse Recombinant FGF-9 (Animal free) (from E. coli)
Mouse Recombinant FGF-9 (Animal free) (from E. coli)
Certificates
undefined
Mouse Recombinant FGF-9 (Animal free) (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    −20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological activity:
    Bioactivity assay: NR6R 3T3 proliferation, ED50 ≤10 ng/ml, bioactivity ≥1.0×10⁵ units/mg
  • Gene ID:
    P54130
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    Heparin-binding growth factor -9 (HBGF-9)|Glia activating factor (GAF)
  • Protein/peptide name:
    FGF-9 (AF)
  • Purity:
    ≥95%
  • Molecular weight:
    23.4 kDa
  • Sequence:
    MPLGEVGSYF GVQDAVPFGN VPVLPVDSPV LLNDHLGQSE AGGLPRGPAV TDLDHLKGIL RRRQLYCRTG FHLEIFPNGT IQGTRKDHSR FGILEFISIA VGLVSIRGVD SGLYLGMNEK GELYGSEKLT QECVFREQFE ENWYNTYSSN LYKHVDTGRR YYVALNKDGT PREGTRTKRH QKFTHFLPRP VDPDKVPELY KDILSQS
  • Endotoxin level:
    Low
  • Formulation:
    10 mM sodium phosphate, pH 7.5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

Specifications

Related Information

Frequently Bought Together