ContactUsLinkComponent
Lincoln-Type Burette Clamp, United Scientific Supplies
Lincoln-Type Burette Clamp, United Scientific Supplies
Catalog # 76293-404
undefined
Lincoln-Type Burette Clamp, United Scientific Supplies
Catalog # 76293-404
Supplier Number:  CLLB02-VWR
Specifications
  • Description:
    Lincoln-Type Burette Clamp
  • Cat. no.:
    76293-404
Specifications
About this item
This zinc-plated stamped steel burette clamp has a steel thumbscrew and can be used with rods up to 3/4" in diameter.
  • Steel thumbscrew
  • Stamped steel
The distance between the two burettes is 7.25" (18.4 cm).