ContactUsLinkComponent
2-Prong Burette Clamp, with Boss Head and Cork-Coated Grips, United Scientific Supplies
2-Prong Burette Clamp, with Boss Head and Cork-Coated Grips, United Scientific Supplies
Catalog # 76293-382
undefined
2-Prong Burette Clamp, with Boss Head and Cork-Coated Grips, United Scientific Supplies
Catalog # 76293-382
Supplier Number:  CLCORK2-VWR
Specifications
  • Description:
    2-Prong Burette Clamp, with Boss Head and Cork-Coated Grips
  • Cat. no.:
    76293-382
Specifications
About this item
The deluxe line of two prong burette clamps are made of pressure die casted zinc alloy and are chrome plated.
  • Corked lined jaws
  • Opens up to 60 mm
Adjustable to hold up to 60 mm O.D. burettes, these clamps feature a spring-type adjustment key. Boss head clamp can swing freely and is lockable at any angle up to 360°. It can be used with rods up to 19 mm.