Order Entry
Canada
ContactUsLinkComponent
Xcelite® Slotted Screwdriver, Apex
Xcelite® Slotted Screwdriver, Apex
Catalog # 76243-992
Supplier:  Cooper Tools
Xcelite® Slotted Screwdriver, Apex
Catalog # 76243-992
Supplier:  Cooper Tools
Supplier Number:  99811N

Specifications

  • Length:
    102 mm (4")
  • Cat. no.:
    76243-992
  • Packaging:
    Boxed
  • Diameter:
    4.76 mm (3/16")

Specifications

About this item