LED Light Units, Production Basics
Catalog # 76253-434
Supplier: Production Basics
Specifications
- Color:Black
- Description:LED Light, with 3 Light Boards and 1 Parts and Hardware Packet
- Dimensions:48"W
- Cat. no.:76253-434
- Bulb Life:50000 h
Specifications
About this item
Direct light for comfort or task with integrated dimmer.
- Philips brand driver and board
- On/Off switch
- Dimmer dial
Features flicker free operation with no hum or buzz.