ContactUsLinkComponent
Saturix Polyurethane Foam Tip Swab with Polypropylene Handle
Saturix Polyurethane Foam Tip Swab with Polypropylene Handle
Catalog # 76240-388
Supplier:  FG CLEAN WIPES TE
Saturix Polyurethane Foam Tip Swab with Polypropylene Handle
Catalog # 76240-388
Supplier:  FG CLEAN WIPES TE
Supplier Number:  6S144
Specifications
  • Material:
    PP handle, PU tip
  • Cat. no.:
    76240-388
  • Packaging:
    50 per bag
  • Handle Length:
    4.5"
  • Tip Width:
    ¹/₄"
Specifications
About this item
These swabs feature polyurethane foam tip heads bonded to polypropylene plastic handles without the use of adhesives.
  • Square-tipped polyurethane foam head
  • Polypropylene plastic handle
  • PU foam head provides good particle entrapment and removal
  • Non sterile
The polyurethane foam head provides good particle entrapment and removal. The absorbent foam enables solvents to be readily sorbed for applications involving solvent delivery or removal. Also ideal for cleaning and applying coatings. The swabs are well suited for cleaning with solvents, with the exception of ketones.