Stripping Tool for Cables, Newark
Catalog # 76253-850
Supplier: NEWARK/ELEMENT 14 TE
Specifications
- Description:Cable Stripping Tool
- Cat. no.:76253-850
- Cutting Capacity:4 - 10 mm
Specifications
About this item
This is a rotary coaxial stripper for use with stripping RG6, RG58 and RG59 cables as well as cable with a diameter between 4 and 10mm
- Rotary and adjustable
- For use with RG6, RG58 and RG59 coaxial cables
The blades can be adjusted to the cable size by inserting an Allen key nto the built-in hex key and turning clockwise for larger cables and anticlockwise for smaller diameter cables.
Delivery: Supplied with Allen key.