Order Entry
Canada
Orders LinkContactUsLinkComponent
Storage Bins, Shaefer Systems

Storage Bins, Shaefer Systems

Storage Bins, Shaefer Systems
Certificates
Storage Bins, Shaefer Systems

3 Options Available Below

Product Details & Documents

Schaefer Shelving complete line of Conductive Containers protect sensitive electronic components from electrostatic fields or induced fields during production, shipping and storage.

  • All containers are returnable, reusable and recyclable
  • Molded with conductive carbon-filled polypropylene
  • Double-wall construction for long term performance
  • Recessed grip handle on back for easier manual handling

Permanently conductive and unaffected by washing with normal cleaners. These containers are molded with a conductive carbon-filled polypropylene material to provide a barrier which these fields cannot penetrate.

Product Family Options

Product Information

  • Color
    Dimensions
    Availability
    Price
  • 76244-978
    EF Stackable Conductive Container Solid Base/Sides
    EF Stackable Conductive Container Solid Base/Sides
    Black
    16W×24L×11H"
    Log in for availability
    Log in for availability
    Each
    Each - $603.23
    $603.23
    Each
       $603.23

    Supplier Number: EF6280.PP3
  • 76248-700
    EF Stackable Conductive Container Solid Base/Sides
    EF Stackable Conductive Container Solid Base/Sides
    Black
    12W×16L×13H"
    Log in for availability
    Log in for availability
    Each
    Each - $341.36
    $341.36
    Each
       $341.36

    Supplier Number: EF4320.PP3

LIDA

Product Information

  • Color
    Dimensions
    Availability
    Price
  • 76244-980
    Stackable Conductive Lid
    Stackable Conductive Lid
    Black
    16W×24L"
    Log in for availability
    Log in for availability
    Each
    Each - $165.53
    $165.53
    Each
       $165.53

    Supplier Number: EFD64.PP3
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.