Corstat® Totes, Conductive Containers
76252-848
- Description:Corstat® in-plant handlers, 20 cells
- Height:5⅜"
- Length:3¾"
- Cat. no.:76252-848
- Width:1¹⁄₁₆"
Used for a standard cell size partition design or other insert.
- Corstat® In-Plant Handlers
Cell length: Will accommodate the dimension of the circuit board that fits left to right in the cell opening. This is the dimension of the edge of the board that will rest on the bottom of the cell.
Cell width: Will accommodate the "profile" of the circuit board. This is the thickness of the board with all of its leads and attached components taken into account.
Cell height (Depth): Will accommodate the dimension of the circuit board up and down as it sits in the cell.