ContactUsLinkComponent
Sewage Pump, Zoro
  76267-958
Sewage Pump, Zoro
  76267-958
 :  WS0511BHF
  • Description:
    Sewage pump
  • Cat. no.:
    76267-958
These Sewage pump includes include vortex impellers that help to eliminate clogging.
They are designed to handle solids-laden wastewater in dewatering, effluent waste handling, onsite wastewater treatment, and septic or municipal sewer system pumping applications.