Orders LinkContactUsLinkComponent
Solid Carbide Square End Mill, Spiral Flute, Sid Tool
Solid Carbide Square End Mill, Spiral Flute, Sid Tool
Square End Mill, 3 Flute, Single End, Solid Carbide, 3/16", ¾", Uncoated, ³⁄₁₆"
Catalog # 76251-098
Supplier:   SID TOOL CO INC TE
Supplier #:  87810487
Solid Carbide Square End Mill, Spiral Flute, Sid Tool
Catalog # 76251-098
Supplier:  SID TOOL CO INC TE
Solid Carbide Square End Mill, Spiral Flute, Sid Tool
Square End Mill, 3 Flute, Single End, Solid Carbide, 3/16", ¾", Uncoated, ³⁄₁₆"
Price
$105.45
Log in for availability
Each
Product Details & Documents
Solid carbide provides better rigidity than high speed steel. It is extremely heat resistant and used for high speed applications on cast iron, nonferrous materials, plastics and other tough-to-machine materials.
  • Uncoated tools do not feature supportive treatments on the cutting edge
  • Used at reduced speeds for general applications on nonferrous metals
  • Uncoated, spiral flute, 40° helix
  • Centercutting with right hand cut
  • Right hand flute, long length
Center cutting tools have one or more cutting edges at the tip and are used in a variety of applications including plunging, drilling or ramping.
Specifications
  • Description:
    Square End Mill, 3 Flute, Single End
  • Length:
    ¾"
  • Material:
    Solid Carbide
  • Finish:
    Uncoated
  • Shank diameter:
    ³⁄₁₆"
  • Diameter:
    3/16"