Orders LinkContactUsLinkComponent
Power Socket Bit Holder, TESTEQUITY LLC TE
Power Socket Bit Holder, TESTEQUITY LLC TE
Bit Holder for 1/4" Drive Power Sockets, 1/4", Black oxide, 1"
Catalog # 76248-196
Supplier:   TESTEQUITY LLC TE
Supplier #:  J6925
Power Socket Bit Holder, TESTEQUITY LLC TE
Catalog # 76248-196
Supplier:  TESTEQUITY LLC TE
Power Socket Bit Holder, TESTEQUITY LLC TE
Bit Holder for 1/4" Drive Power Sockets, 1/4", Black oxide, 1"
Price
$37.30
Log in for availability
Each
Product Details & Documents
A Proto® power socket bit holder for 1/4" drives. Not for use in impact applications. Black oxide finish prevents corrosion.
Specifications
  • Description:
    Bit Holder for 1/4" Drive Power Sockets
  • Overall length:
    1"
  • Finish:
    Black oxide
  • Drive size:
    1/4"