Order Entry
Canada
Orders LinkContactUsLinkComponent
Socket Adapter, Sid Tool Co

Socket Adapter, Sid Tool Co

Socket Adapter, Drive Size 3/8", Drive - Female Square, Impact Drills, Cordless Drills, Corded Drills, Ratchets, 1.9×0.7×3.5", 0.32 oz
Catalog # 76267-782
Supplier:   SID TOOL CO INC TE
Supplier #:  43966944
Socket Adapter, Sid Tool Co
Catalog # 76267-782
Supplier:  SID TOOL CO INC TE
Socket Adapter, Sid Tool Co

Socket Adapter, Drive Size 3/8", Drive - Female Square, Impact Drills, Cordless Drills, Corded Drills, Ratchets, 1.9×0.7×3.5", 0.32 oz

Price
167,90 $
Log in for availability
Each
Product Details & Documents

Performance Series Socket Reducers make smaller square drive sockets and accessories compatible with 3/8" or 1/2" square drive drills or accessories.

  • Built for maximum durability and long life with impact drivers
  • Black oxide finish resists corrosion and increases life

Manufactured with heavy-duty, cold-forged, high-grade steel to withstand high torque output.

Specifications
  • Description:
    Socket Adapter, Drive Size 3/8"
  • Dimensions:
    1.9×0.7×3.5"
  • For use with:
    Impact Drills, Cordless Drills, Corded Drills, Ratchets
  • Type:
    Drive - Female Square
  • Weight:
    0.32 oz
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.