Precision Potentiometer, Digi-Key
Precision Potentiometer
Catalog # 76251-628
Supplier: DIGI-KEY CORPORATION
Supplier #: 3590S-2-103L
Jump to:
Product Details & Documents
Precision Potentiometer with plastic or metal shaft and bushings.
- Wirewound
- Solder lugs or PC pins
- Sealable (Full body seal)
Designed for use in HMI applications and with optional AR pin feature.
: RoHS compliant.
Specifications
- Description:Precision Potentiometer