Orders LinkContactUsLinkComponent
Taper Shank, CAT40, Sid Tool Co
Taper Shank, CAT40, Sid Tool Co
Taper Shank, CAT40, 1/8" hole I.D., 5/8 - 11 thread
Catalog # 76253-136
Supplier:   SID TOOL CO INC TE
Supplier #:  80263767
Taper Shank, CAT40, Sid Tool Co
Catalog # 76253-136
Supplier:  SID TOOL CO INC TE
Taper Shank, CAT40, Sid Tool Co
Taper Shank, CAT40, 1/8" hole I.D., 5/8 - 11 thread
Price
$863.46
Log in for availability
Each
Product Details & Documents
CAT40 Taper Shank, with end mill holder.
  • DIN flange coolant through
  • Through-spindle coolant
  • Maximum TIR 0.0002″ OD to ID
Shank traverse ground to obtain optimum accuracy.
Certifications: Meet or exceed ANSI specifications.
Delivery information: Each tool comes with inspection lab certificate.
Specifications
  • Description:
    Taper Shank, CAT40, 1/8" hole I.D., 5/8 - 11 thread