TorqueControl Square Drive Adapter Blade, Wiha Tools
Catalog # 76251-684
Supplier: WIHA TOOLS TE
Specifications
- Description:Square Drive Adapter Torque Control Blade, Head type - Adapter
- Cat. no.:76251-684
- Weight:0.0800 lbs.
- Drive Size:For 1/4" Sockets
Specifications
About this item
TorqueControl Square Drive Adapter Blade.
- Adapter fits all 1/4" Sockets
- Corrosion protective Hard Chrome Finish
Wiha special chrome-vanadium-molybdenum tool steel, through hardened for best wear resistance. Not for use in wiha insulated torque control handles.
Certifications: RoHS Compliant.