ESD Floor Mat Grounding Kit, Transforming Technologies
76250-302
- Color:Black
- Description:Floor Mat Ground Kit
- Cat. no.:76250-302
This ground Kit comes with a molded low profile ground cord with mushroom cap and a 3/8 (10 mm) snap ensures secure connection to floor mats.
: Meets or exceeds requirements of ANSI ESD-S20.20 and ANSI/ESD S-6.1.