Shielding Bag, SCS Division of Desco, Desco Industries
Catalog # 76249-724
Supplier: SCS DIVISION OF DESCO TE
Specifications
- Dimensions:Metal-In static shielding bag
- Size:15×18"
- Cat. no.:76249-724
Specifications
About this item
Static Shield Bag 81705 Series are manufactured from four layers static dissipative coating, polyester, metal and polyethylene laminate.
- The polyester dielectric in concert with the metal layer provides discharge shielding
The exterior is static dissipative and allows electrostatic charges to be removed when grounded.