Tip Detector, EFD
Tip Detector Accessory Kit, EV Series
Catalog # 76249-810
Supplier: EFD INC.- CUSTOMER SERVICE TE
Supplier #: 7360893
Jump to:
Product Details & Documents
Tip detector allows to automatically update both the XY offsets and the Z height by clicking on needle Z detect in the EV series automated dispenser systems.
- The needle Z detect button is present only on systems that include the tip detector
Specifications
- Description:Tip Detector Accessory Kit, EV Series