Soder-Wick® Rosin Flux Desoldering Braid, ITW Chemtronics
Catalog # 76243-440
Supplier: ITW Chemtronics
Specifications
- Width:0.145"
- Color:Brown
- Description:Soder-Wick® Rosin Flux Desoldering Braid on Spool
- Length:25'
- Size:#5
- Cat. no.:76243-440
Specifications
About this item
Soder-Wick® Rosin Flux Desoldering Braid is designed for heat sensitive electronic components using lighter mass, pure copper braid construction that allows for better thermal conductivity, even at low temperatures.
- Noncorrosive ultra-high purity Type R rosin flux
- Minimizes the risk of heat damage to the board
- Will not leave ionic contamination on the boards
Minimizes the risk of damage associated with static electricity. This braid responds faster than conventional desoldering braids thereby minimizing overheating and preventing PCB damage.
Certifications: Meets MIL-F-14256 F, DOD-STD-883E, Method 2022, ANSI/IPC J STD-004 type ROLO and NASA-STD-8739.3 Soldered Electrical Connections.
Packaging: Spools packaged in ESD-safe static dissipative bobbins. Sealed in nitrogen-purged packaging to avoid corrosion and loss of performance from moisture and oxygen.