Thomas and Betts Ratchet Crimp Tool, Newark/Element
Catalog # 76253-376
Supplier: NEWARK/ELEMENT 14 TE
Specifications
- Description:Ratchet Hand Crimp Tool
- Cat. no.:76253-376
Specifications
About this item
This is a manual crimp tool for Shield-kon non-insulated outer sleeve GSC connectors and insulated outer sleeve GSR connectors.
- MIL-specified parallel-action hand tool with Shure-Stake mechanism
Delivery: Supplied in a wooden box containing one frame.